Product Details
- SNP ID
-
rs3692583
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:16605397 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCATCCCAGCACCAACATTTCCTAT[G/T]CATCCCCTGCTTCTAACAGAGCTTG
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Hgf
PubMed Links
Gene Details
- Gene
- Hgf
- Gene Name
- hepatocyte growth factor
There are no transcripts associated with this gene.
View Full Product Details