Product Details

SNP ID
rs4228647
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:32230040 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TCTATGGCATGTACTTCTGCTTGAA[T/C]GTCTCTAGTCAAGGTTAGTGGTGCT
Phenotype
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
Mpg PubMed Links

Gene Details

Gene
Mpg
Gene Name
N-methylpurine-DNA glycosylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_010822.3 603 Silent Mutation NP_034952.2
Gene
Nprl3
Gene Name
nitrogen permease regulator-like 3
There are no transcripts associated with this gene.

Gene
Rhbdf1
Gene Name
rhomboid family 1 (Drosophila)
There are no transcripts associated with this gene.

View Full Product Details