Product Details
- SNP ID
-
rs13476608
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:82994075 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTAAGAATCACAACAATGGGGGAGA[C/T]GGGCAAATAGCAAGCCAGTTCTTTG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Fsip2
PubMed Links
Gene Details
- Gene
- Fsip2
- Gene Name
- fibrous sheath-interacting protein 2
There are no transcripts associated with this gene.
View Full Product Details