Product Details
- SNP ID
-
rs4228162
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:21125250 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAAGAAGTGCAATGCTTTTATAAA[C/T]CGACAATAACAACAAAAAAAAAAAA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Myb
PubMed Links
Gene Details
- Gene
- Myb
- Gene Name
- myeloblastosis oncogene
View Full Product Details