Product Details

SNP ID
rs13475697
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:3395746 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CCATAGTTTGAGTTCTCCCAAAATA[G/T]CAAATGTCAGCATCAGCCAAATAAA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
Xkr4 PubMed Links

Gene Details

Gene
Xkr4
Gene Name
X Kell blood group precursor related family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001011874.1 Intron NP_001011874.1

View Full Product Details