Product Details

SNP ID
rs13464858
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.13:108279645 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CGTGGTCATGTATTCTTACTATGGG[C/T]TATGTGCAATGGGACCAGCCTACCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Elovl7 PubMed Links

Gene Details

Gene
Elovl7
Gene Name
ELOVL family member 7, elongation of long chain fatty acids (yeast)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_029001.5 733 Silent Mutation NP_083277.3

View Full Product Details