Product Details

SNP ID
rs13480505
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.10:14143850 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CTCTCCCAGAAGAGAGAGAAGATAT[A/C]TGACCACCATCAGAGCACCGTCTCC
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
Hivep2 PubMed Links

Gene Details

Gene
Hivep2
Gene Name
human immunodeficiency virus type I enhancer binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_010437.2 7197 Missense Mutation NP_034567.2

View Full Product Details