Product Details

SNP ID
rs13480894
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:17945698 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
ATGAATTGACAGTTTCCCTTTTTCC[C/T]AAATCCTCTGAGATTTTGCCAACAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Etaa1 PubMed Links

Gene Details

Gene
Etaa1
Gene Name
Ewing tumor-associated antigen 1
There are no transcripts associated with this gene.

Gene
Etaa1os
Gene Name
Ewing tumor-associated antigen 1, opposite strand
There are no transcripts associated with this gene.

View Full Product Details