Product Details

SNP ID
rs6370720
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.6:56357276 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
ATTCCATGTTGCTTCCTACTCCATG[C/T]CTTCTTCAGCATCATCTCTCACTCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Pde1c PubMed Links

Gene Details

Gene
Pde1c
Gene Name
phosphodiesterase 1C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025568.2 Intron NP_001020739.1
NM_001159952.1 Intron NP_001153424.1
NM_001159953.1 Intron NP_001153425.1
NM_001159955.1 Intron NP_001153427.1
NM_001159956.1 Intron NP_001153428.1
NM_001159957.1 Intron NP_001153429.1
NM_001159960.1 Intron NP_001153432.1
NM_011054.4 Intron NP_035184.1

View Full Product Details