Product Details

SNP ID
rs6244569
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.14:28908015 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CAGGATGGAAGCCGTGGCAGGATTC[C/T]GGACGTCGTCTGATCTTCTGAGCCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Cacna2d3 PubMed Links

Gene Details

Gene
Cacna2d3
Gene Name
calcium channel, voltage-dependent, alpha2/delta subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_009785.1 3253 Silent Mutation NP_033915.1

View Full Product Details