Product Details
- SNP ID
-
rs13482120
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:30613538 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACACACTGCATAGCCTTGTAATTAT[A/G]TTAGCTCACTTTGTCCCAGTGTAGC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Prkcd
PubMed Links
Gene Details
- Gene
- Prkcd
- Gene Name
- protein kinase C, delta
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_011103.3 |
|
Intron |
|
|
NP_035233.1 |
View Full Product Details