Product Details

SNP ID
rs4151923
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.16:13814453 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TGGTGGCAAGTCACTAGGAAACTTC[A/C]AGGTCTGTCTCATCTTTATTTTACT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
Ntan1 PubMed Links

Gene Details

Gene
Ntan1
Gene Name
N-terminal Asn amidase
There are no transcripts associated with this gene.

Gene
Rrn3
Gene Name
RRN3 RNA polymerase I transcription factor homolog (yeast)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039521.1 3210 UTR 3 NP_001034610.1

View Full Product Details