Product Details
- SNP ID
-
rs6346451
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:141680716 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCTAAGCAATACTCCTAGAAATGA[G/T]GCTGACCTCTACTTCTTCTGCCCTT
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Macrod2
PubMed Links
Gene Details
- Gene
- Macrod2
- Gene Name
- MACRO domain containing 2
View Full Product Details