Product Details

SNP ID
rs3681171
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.6:18108468 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GGTCAGTGCTTTCTTAAATGTCTCG[T/G]TCTTCTCTTCAACGGGTAAAGACCT
Phenotype
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
Asz1 PubMed Links

Gene Details

Gene
Asz1
Gene Name
ankyrin repeat, SAM and basic leucine zipper domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_023729.3 165 Missense Mutation NP_076218.3

View Full Product Details