Product Details

SNP ID
rs13481059
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:62373387 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CTACGTCCATCGGCTCTGCGGTCTC[A/G]GCACTGGCGCTGTCGGTCACCTGGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Ncor1 PubMed Links

Gene Details

Gene
Ncor1
Gene Name
nuclear receptor co-repressor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252313.1 2488 Silent Mutation NP_001239242.1
NM_011308.3 2488 Silent Mutation NP_035438.3
NM_177229.3 2488 Intron NP_796203.1

View Full Product Details