Product Details

SNP ID
rs13476817
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.2:144373653 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TCAGTCGGCATGACGATGACGCCAC[A/G]AGAACGTCCACCTCGGAAGGGCTGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Csrp2bp PubMed Links

Gene Details

Gene
Csrp2bp
Gene Name
cysteine and glycine-rich protein 2 binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166640.1 586 Intron NP_001160112.1
NM_181417.3 586 Silent Mutation NP_852082.2
Gene
Pet117
Gene Name
PET117 homolog (S. cerevisiae)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164813.1 586 Intron NP_001158285.1

View Full Product Details