Product Details

SNP ID
rs13477392
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.3:127540833 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CCTCGAGGTTCCAACTGTGCTTCTT[C/T]CTAATTTCAGTCTGTGCATTCATTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Larp7 PubMed Links

Gene Details

Gene
Larp7
Gene Name
La ribonucleoprotein domain family, member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138593.2 1674 Silent Mutation NP_613059.2
Gene
Mir302a
Gene Name
microRNA 302a
There are no transcripts associated with this gene.

Gene
Mir302b
Gene Name
microRNA 302b
There are no transcripts associated with this gene.

Gene
Mir302c
Gene Name
microRNA 302c
There are no transcripts associated with this gene.

Gene
Mir302d
Gene Name
microRNA 302d
There are no transcripts associated with this gene.

Gene
Mir367
Gene Name
microRNA 367
There are no transcripts associated with this gene.

View Full Product Details