Product Details
- SNP ID
-
rs6369115
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:84443271 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATACATGGAAGGAATAATGCATTG[C/T]GTTAAAGCAATAATCAGAAGCCTGT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Bnc2
PubMed Links
Gene Details
- Gene
- Bnc2
- Gene Name
- basonuclin 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_172870.4 |
|
Intron |
|
|
NP_766458.3 |
View Full Product Details