Product Details
- SNP ID
-
rs13476265
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:179610525 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGATGCTGAAGATTCCCTGTCCTA[C/T]GAGGACATCCAGGACGACGACTCTG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Cnst
PubMed Links
Gene Details
- Gene
- Cnst
- Gene Name
- consortin, connexin sorting protein
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_146105.3 |
1793 |
Silent Mutation |
|
|
NP_666217.2 |
View Full Product Details