Product Details
- SNP ID
-
rs4197745
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:67589257 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCATCAATGGGTGAATACGAATGCA[C/T]CCTACTGAGGGATTTTTACAAAACG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Cadm2
PubMed Links
Gene Details
- Gene
- Cadm2
- Gene Name
- cell adhesion molecule 2
View Full Product Details