Product Details

SNP ID
rs13481061
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.11:62992617 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TGATACTGCTTGAAAAACACAACCC[G/T]TGGCCAGCCTACGTCGCTTACATCT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
Cdrt4 PubMed Links

Gene Details

Gene
Cdrt4
Gene Name
CMT1A duplicated region transcript 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025496.1 245 Silent Mutation NP_079772.1
Gene
Cdrt4os1
Gene Name
CMT1A duplicated region transcript 4, opposite strand 1
There are no transcripts associated with this gene.

View Full Product Details