Product Details

SNP ID
rs13475755
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:20935145 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GCTCCGACCAGGCCTGGTATGAGCT[A/G]TTTTGGATTTTCTTCCTGCCGGCGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Paqr8 PubMed Links

Gene Details

Gene
Paqr8
Gene Name
progestin and adipoQ receptor family member VIII
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_028829.3 809 Silent Mutation NP_083105.3

View Full Product Details