Product Details

SNP ID
rs13475765
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:23815235 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CCCTTTTGTAGATGAGGACCACTCG[C/G]AAGGTTTCTGTCTGGCCTGTGGGAC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
B3gat2 PubMed Links

Gene Details

Gene
B3gat2
Gene Name
beta-1,3-glucuronyltransferase 2 (glucuronosyltransferase S)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172124.2 1318 UTR 3 NP_742122.2

View Full Product Details