Product Details

SNP ID
rs13478106
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.5:8421631 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GAGCTGAATCAGCAGAAAATAGTTA[C/T]TATTTACTCCTGTTCCGTGTATGAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Dbf4 PubMed Links

Gene Details

Gene
Dbf4
Gene Name
DBF4 homolog (S. cerevisiae)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190717.1 Intron NP_001177646.1
NM_013726.3 Intron NP_038754.1
Gene
Slc25a40
Gene Name
solute carrier family 25, member 40
There are no transcripts associated with this gene.

View Full Product Details