Product Details

SNP ID
rs13481471
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.12:56901420 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
GAGTTGAGCTGGTCCTCCAGGGATA[A/T]AAGAACTGCCCCTGGGAAGAGCTCC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
Slc25a21 PubMed Links

Gene Details

Gene
Slc25a21
Gene Name
solute carrier family 25 (mitochondrial oxodicarboxylate carrier), member 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167976.1 Intron NP_001161448.1
NM_172577.3 Intron NP_766165.2

View Full Product Details