Product Details

SNP ID
rs6351323
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.3:32464232 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TAACCCATTTTACTATCATGTCCCA[A/T]TTCCAAGTGGTGAGGTCTCACTCCA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
Kcnmb3 PubMed Links

Gene Details

Gene
Kcnmb3
Gene Name
potassium large conductance calcium-activated channel, subfamily M, beta member 3
There are no transcripts associated with this gene.

Gene
Pik3ca
Gene Name
phosphatidylinositol 3-kinase, catalytic, alpha polypeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_008839.2 4663 UTR 3 NP_032865.2

View Full Product Details