Product Details

SNP ID
rs6248251
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:123438077 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TTTATTGCACACTTGTGATCAGTCT[A/G]ATCAGAAAAAAAATTCCTAAAGACA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Dpp10 PubMed Links

Gene Details

Gene
Dpp10
Gene Name
dipeptidylpeptidase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_199021.3 Intron NP_950186.3

View Full Product Details