Product Details
- SNP ID
-
rs13481423
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:44477843 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCACATTTTTAATTTCCCACATGCT[A/T]GGAACAGTGTCAAAATTAAATGGTA
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Nrcam
PubMed Links
Gene Details
- Gene
- Nrcam
- Gene Name
- neuronal cell adhesion molecule
View Full Product Details