Product Details
- SNP ID
-
rs13459154
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:65861451 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGTCTTCCCACAAGCCTCTTCAAA[C/T]CGGGGCATAAGCTCTTTGGTAAGCA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Ralbp1
PubMed Links
Gene Details
- Gene
- Ralbp1
- Gene Name
- ralA binding protein 1
View Full Product Details