Product Details
- SNP ID
-
rs4222538
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:93022561 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGGTGAGTGAGGGTACCAGCCAGG[A/G]CAGGTGCTGGCTGCCGTGCGCGGCA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Kif1a
PubMed Links
Gene Details
- Gene
- Kif1a
- Gene Name
- kinesin family member 1A
There are no transcripts associated with this gene.
View Full Product Details