Product Details
- SNP ID
-
rs13482888
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:14923629 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTACATAAACAGGCTGAGAAACACT[A/G]TATGTCCTAACTAAGCTGCAAATGG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Wdr27
PubMed Links
Gene Details
- Gene
- Wdr27
- Gene Name
- WD repeat domain 27
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_175173.3 |
|
Intron |
|
|
NP_780382.2 |
View Full Product Details