Product Details
- SNP ID
-
rs13477128
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:57437613 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGACCAAGCCTTGTGGAGCGAGTGC[A/G]AAGAGCCCCGTGATGTGGTCCCCTG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Tm4sf4
PubMed Links
Gene Details
- Gene
- Tm4sf4
- Gene Name
- transmembrane 4 superfamily member 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_145539.2 |
644 |
Missense Mutation |
|
|
NP_663514.2 |
View Full Product Details