Product Details

SNP ID
rs13459138
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.12:112821840 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
ACAGGATCTCTGTGGACACCAGGCG[C/T]TCAACTGGTGCCCGATCTACATTCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BC022687 PubMed Links

Gene Details

Gene
BC022687
Gene Name
cDNA sequence BC022687
There are no transcripts associated with this gene.

Gene
Cdca4
Gene Name
cell division cycle associated 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_028023.3 814 Silent Mutation NP_082299.1

View Full Product Details