Product Details
- SNP ID
-
rs13475870
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:51786298 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAGGAGAGAGAGGAAAATGACTCG[A/T]GTCCTTGGATACATGATGTGCTCTG
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Myo1b
PubMed Links
Gene Details
- Gene
- Myo1b
- Gene Name
- myosin IB
There are no transcripts associated with this gene.
View Full Product Details