Product Details
- SNP ID
-
rs13476215
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:164411839 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAACGTCAAAAGATTTTCCTGTTC[A/G]TGTGTCCAATGCCTCCAACACCGTG
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
Nme7
PubMed Links
Gene Details
- Gene
- Nme7
- Gene Name
- NME/NM23 family member 7
There are no transcripts associated with this gene.
View Full Product Details