Product Details
- SNP ID
-
rs3672870
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:73583765 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGGTTTAGCCTTAACAGTTATGAA[G/C]TCTGTAGAGTAGTGATCTATAAAAC
- Phenotype
-
- Polymorphism
- G/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Slc45a4
PubMed Links
Gene Details
- Gene
- Slc45a4
- Gene Name
- solute carrier family 45, member 4
View Full Product Details