Product Details
- SNP ID
-
rs6298689
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:139764653 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGGCCGTGCTCCCTCCAGCACTCC[C/T]GGGGGAGGACTCACCTGGCGCTGGA
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Ints1
PubMed Links
Gene Details
- Gene
- Ints1
- Gene Name
- integrator complex subunit 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_026748.2 |
|
Intron |
|
|
NP_081024.3 |
- Gene
- Mir7037
- Gene Name
- microRNA 7037
There are no transcripts associated with this gene.
View Full Product Details