Product Details

SNP ID
rs3690198
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.13:60718040 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CGGAAAACATAATGCTTCTGGATAG[A/G]AATGTGCCCAAACCTCGGATCAAGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Dapk1 PubMed Links

Gene Details

Gene
Dapk1
Gene Name
death associated protein kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001285917.1 609 Silent Mutation NP_001272846.1
NM_029653.3 609 Silent Mutation NP_083929.2
NM_134062.2 609 Silent Mutation NP_598823.1

View Full Product Details