Product Details
- SNP ID
-
rs13476968
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:9280970 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGCCCGATGAGGAGCTGGCTGAAGA[C/T]GGACAAGATGAAAACGATGCACCTC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Zbtb10
PubMed Links
Gene Details
- Gene
- Zbtb10
- Gene Name
- zinc finger and BTB domain containing 10
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_177660.3 |
3086 |
Silent Mutation |
|
|
NP_808328.2 |
View Full Product Details