Product Details
- SNP ID
-
rs13477205
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:76233869 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGATCAAGCAGGGTTTCTTACAAAC[C/G]TTTATAACCATGAGCAATTTCTTTA
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Fstl5
PubMed Links
Gene Details
- Gene
- Fstl5
- Gene Name
- follistatin-like 5
View Full Product Details