Product Details
- SNP ID
-
rs13483812
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:69834798 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCAACAGCAACAACTTATCCACCA[A/G]CAATGAGGAGACAGCTTTTTCCCCG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Aff2
PubMed Links
Gene Details
- Gene
- Aff2
- Gene Name
- AF4/FMR2 family, member 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_008032.3 |
2650 |
Missense Mutation |
|
|
NP_032058.2 |
View Full Product Details