Product Details
- SNP ID
-
rs3674280
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:153233346 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGTAAACACTCTAATTTTGAAGTG[G/A]AAGAAAAAGAGCATGCATATCTGGT
- Phenotype
-
- Polymorphism
- G/A, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Lamc1
PubMed Links
Gene Details
- Gene
- Lamc1
- Gene Name
- laminin, gamma 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_010683.2 |
|
Intron |
|
|
NP_034813.2 |
View Full Product Details