Product Details
- SNP ID
-
rs4187387
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:55380886 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATGGGAGATAAAGAAACATTAAGC[A/T]CAGTTTCCTCTGGGAAGTGGAATTA
- Phenotype
-
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Zpld1
PubMed Links
Gene Details
- Gene
- Zpld1
- Gene Name
- zona pellucida like domain containing 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_178720.4 |
|
Intron |
|
|
NP_848835.1 |
View Full Product Details