Product Details

SNP ID
rs13480477
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.10:6974370 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CAATCAATCAAGATAGTTCTCTTAT[T/C]GAGAAATTCTGGCTTTCAAGTAAGA
Phenotype
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
Ipcef1 PubMed Links

Gene Details

Gene
Ipcef1
Gene Name
interaction protein for cytohesin exchange factors 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033391.2 Intron NP_001028563.1
NM_001170800.1 Intron NP_001164271.1
NM_001170801.1 Intron NP_001164272.1
NM_001170802.1 Intron NP_001164273.1
Gene
Oprm1
Gene Name
opioid receptor, mu 1
There are no transcripts associated with this gene.

View Full Product Details