Product Details

SNP ID
rs4165334
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.16:23467605 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TACAGCCCAACCTTTGCACCACTGA[C/T]CTGGGCCTAAGCTCCCTTGTCTGCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Masp1 PubMed Links

Gene Details

Gene
Masp1
Gene Name
mannan-binding lectin serine peptidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_008555.2 Intron NP_032581.2

View Full Product Details