Product Details

SNP ID
rs6316191
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:11271646 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
TGATCTGATAAACAGATAAATAACA[C/T]AGGTGTATGTAATTTCTAAAAGTAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
Prex2 PubMed Links

Gene Details

Gene
Prex2
Gene Name
phosphatidylinositol-3,4,5-trisphosphate-dependent Rac exchange factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033636.4 Intron NP_001028808.1
NM_029525.1 Intron NP_083801.1

View Full Product Details