Product Details
- SNP ID
-
rs13481941
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:91537060 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCATTTCTCCCATTCTTCATCATTA[A/C]AGCTTTACCTACAACAGAAGGATCA
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
Ssbp2
PubMed Links
Gene Details
- Gene
- Ssbp2
- Gene Name
- single-stranded DNA binding protein 2
View Full Product Details