Product Details

SNP ID
rs13475864
Assay Type
Pre Designed
NCBI dbSNP Submissions
NA
Location
Chr.1:51064876 on Build GRCm38
Set Membership
Context Sequence [VIC/FAM]
CAAAGTGTGGCATCATAGAAATATA[A/G]GTCAGACAGCCATGACATGAGGCAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
Tmeff2 PubMed Links

Gene Details

Gene
Tmeff2
Gene Name
transmembrane protein with EGF-like and two follistatin-like domains 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019790.4 Intron NP_062764.1

View Full Product Details