Product Details
- SNP ID
-
rs13467342
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:32718584 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTCTCAGGAGGCTGTTCGAGAAGG[C/T]TCTCCAGCCAACTGGAAAAGAAAAG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Actl6a
PubMed Links
Gene Details
- Gene
- Actl6a
- Gene Name
- actin-like 6A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_019673.2 |
1006 |
Silent Mutation |
|
|
NP_062647.2 |
- Gene
- Mrpl47
- Gene Name
- mitochondrial ribosomal protein L47
There are no transcripts associated with this gene.
View Full Product Details