Product Details
- SNP ID
-
rs6378225
- Assay Type
- Pre Designed
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:8591730 on Build GRCm38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTACTGAGATGTTGATTACAGAAGA[A/G]TCAACTCCTCTGTCCAGCTGATACT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
Glcci1
PubMed Links
Gene Details
- Gene
- Glcci1
- Gene Name
- glucocorticoid induced transcript 1
View Full Product Details